Anti-TRIM17 Rabbit Recombinant Polyclonal Antibody
Catalog # 77846-556
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TRIM17
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y577
- Antigen synonyms:E3 ubiquitin-protein ligase TRIM17|RBCC|RING finger protein 16|RING finger protein terf|RNF16|TERF|Testis RING finger protein|TRI17_HUMAN|TRIM 17|Trim17|Tripartite motif containing 17|Tripartite motif containing protein 17|Tripartite motif protein 17|Tripartite motif-containing protein 17
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human E3 ubiquitin-protein ligase TRIM17 protein (1-300AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: TRIM17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human