Anti-CSF2RB Rabbit Recombinant Polyclonal Antibody
77853-650
New Product
- Antigen symbol:CSF2RB
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P32927
- Antigen synonyms:Beta-C|CD131|CDw131|Colony stimulating factor 2 receptor beta|Colony stimulating factor 2 receptor beta low affinity (granulocyte macrophage)|Common beta chain|Cytokine receptor common beta chain|Cytokine receptor common beta chain precursor (CDw131 antigen)|Cytokine receptor common subunit beta|GM CSF/IL 3/IL 5 receptor common beta chain|GM-CSF/IL-3/IL-5 receptor common beta subunit|granulocyte-macrophage colony-stimulating factor receptor|beta|IL3RB|IL3RB_HUMAN|IL5RB|Interleukin 3 receptor|Interleukin 3 receptor/granulocyte macrophage colony stimulating factor 3 receptor beta (high affinity)|Interleukin 5 receptor|SMDP5|stimulating factor 3 receptor|beta (high affinity)
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cytokine receptor common subunit beta protein (492-710AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CSF2RB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human