Anti-ERLIN2 Rabbit Recombinant Polyclonal Antibody
77854-901
New Product
- Antigen symbol:ERLIN2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O94905
- Antigen synonyms:C8orf2|Chromosome 8 open reading frame 2|Endoplasmic reticulum lipid raft associated protein 2|Endoplasmic reticulum lipid raft-associated protein 2|ER lipid raft associated 2|ERLIN 2|Erlin-2|ERLN2_HUMAN|HGNC:1356|MGC87072|NET32|SPFH 2|SPFH domain containing protein 2|SPFH domain family member 2|SPFH domain-containing protein 2|SPFH2|SPG18|Stomatin prohibitin flotillin HflC/K domain containing protein 2|Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human Erlin-2 protein (212-339AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ERLIN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human