Anti-ACTL7B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACTL7B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-634
- 77846-635
- Antigen symbol:
- ACTL7B
- ACTL7B
- Antigen synonyms:
- ACL7B_HUMAN|Actin like 7 beta|Actin like 7B|Actin like protein 7B|Actin-like protein 7B|Actin-like-7-beta|MGC151192|MGC151194|OTTHUMP00000021867|OTTMUSP00000007742|RP11-3J11.2|RP23-48F4.1|t actin 1|Tact1
- ACL7B_HUMAN|Actin like 7 beta|Actin like 7B|Actin like protein 7B|Actin-like protein 7B|Actin-like-7-beta|MGC151192|MGC151194|OTTHUMP00000021867|OTTMUSP00000007742|RP11-3J11.2|RP23-48F4.1|t actin 1|Tact1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y614
- Q9Y614
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Actin-like protein 7B protein (1-415AA)
- Recombinant Human Actin-like protein 7B protein (1-415AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together