Anti-ENTPD2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-759
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ENTPD2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9Y5L3
- Antigen synonyms:NTPDase 2|NTPDase2|CD39 antigen like 1|CD39 antigen-like 1|CD39 like1|CD39L1|CD39like1|ecto ATP diphosphohydrolase 2|ecto ATPase 2|ecto ATPDase 2|Ecto-ATP diphosphohydrolase 2|Ecto-ATPase 2|Ecto-ATPDase 2|ectoATPase 2|ectoATPDase 2|Ectonucleoside triphosphate diphosphohydrolase 2|ENTP2_HUMAN|Entpd2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 2 protein (28-268AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ENTPD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human