Anti-DMKN Rabbit Recombinant Polyclonal Antibody
77854-229
New Product
- Antigen symbol:DMKN
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q6E0U4
- Antigen synonyms:Dermokine|DMKN_HUMAN|Epidermis-specific secreted protein SK30/SK89|FLJ79374|UNQ729|UNQ729/PRO1411|ZD52F10
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dermokine protein (1-137AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: DMKN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse