Anti-CHEK2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-053
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CHEK2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O96017
- Antigen synonyms:CDS 1|Cds1|Cds1 homolog|Checkpoint kinase 2|Checkpoint like protein CHK2|CHEK 2|Chk 2|CHK2 checkpoint homolog (S. pombe)|CHK2 checkpoint homolog|CHK2_HUMAN|hCds1|HuCds 1|LFS 2|LFS2|PP1425|RAD 53|RAD53|Rad53 homolog|Serine/threonine protein kinase Chk2|Serine/threonine-protein kinase Chk2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Serine/threonine-protein kinase Chk2 protein (1-270AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: CHEK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human