Bovine Recombinant Phosphodiesterase (from Spodoptera frugiperda)
Catalog # CA80059-350
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Enzyme type:Recombinant
- Source:Spodoptera frugiperda
- Species:Bovine
- Size:50 μl
- Storage conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Enzyme name:Phosphodiesterase
- Enzyme synonyms:PDE V;Phosphodiesterase V;cGMP-binding cGMP-specific PDE