Anti-WASF3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WASF3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-178
- 77844-177
- Antigen symbol:
- WASF3
- WASF3
- Antigen synonyms:
- KIAA0900|Protein WAVE-3|Protein WAVE3|SCAR3|Verprolin homology domain containing protein 3|Verprolin homology domain-containing protein 3|WASF3_HUMAN|WASP family protein member 3|WAVE3|Wiskott Aldrich syndrome protein family member 3|Wiskott-Aldrich syndrome protein family member 3
- KIAA0900|Protein WAVE-3|Protein WAVE3|SCAR3|Verprolin homology domain containing protein 3|Verprolin homology domain-containing protein 3|WASF3_HUMAN|WASP family protein member 3|WAVE3|Wiskott Aldrich syndrome protein family member 3|Wiskott-Aldrich syndrome protein family member 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPY6
- Q9UPY6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Wiskott-Aldrich syndrome protein family member 3 protein (368-502AA)
- Recombinant Human Wiskott-Aldrich syndrome protein family member 3 protein (368-502AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together