Anti-TBL1X Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: TBL1X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-851
- 77847-850
- Antigen symbol:
- TBL1X
- TBL1X
- Antigen synonyms:
- EBI|F box like/WD repeat protein F-box-like/WD repeat-containing protein SMAP 55|SMAP55|TBL 1|TBL 1X|TBL1|TBL1X_HUMAN|Transducin (beta) like 1|Transducin (beta) like 1 X linked|Transducin (beta) like 1X linked|Transducin beta like 1 X linked|Transducin beta like 1X|Transducin beta like 1X protein|Transducin beta like protein 1|X linked|Transducin beta-like protein 1X|Transducin-beta-like protein 1|X-linked
- EBI|F box like/WD repeat protein F-box-like/WD repeat-containing protein SMAP 55|SMAP55|TBL 1|TBL 1X|TBL1|TBL1X_HUMAN|Transducin (beta) like 1|Transducin (beta) like 1 X linked|Transducin (beta) like 1X linked|Transducin beta like 1 X linked|Transducin beta like 1X|Transducin beta like 1X protein|Transducin beta like protein 1|X linked|Transducin beta-like protein 1X|Transducin-beta-like protein 1|X-linked
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60907
- O60907
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TBL1X
- Fusion protein of Human TBL1X
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together