Anti-ANGPTL2 Rabbit Recombinant Polyclonal Antibody
77844-247
New Product
- Antigen symbol:ANGPTL2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:Q9UKU9
- Antigen synonyms:AI593246|Angiopoietin like 2|Angiopoietin related protein 2|Angiopoietin-like protein 2|Angiopoietin-related protein 2|ANGL2_HUMAN|Angptl2|Arp2|AW260363|HARP|MGC8889|UNQ170/PRO196
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Angiopoietin-related protein 2 protein (23-493AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:500 to 1:2000, IHC:1:20 to 1:200
Type:
Antigen: ANGPTL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse