Anti-CKS2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-172
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CKS2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P33552
- Antigen synonyms:CDC28|CDC28 protein kinase 2|CDC28 protein kinase regulatory subunit 2|CKS 2|CKS-2|CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog 2|CKS2_HUMAN|CKSHS2|Cyclin dependent kinases regulatory subunit 2|Cyclin-dependent kinases regulatory subunit 2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cyclin-dependent kinases regulatory subunit 2 protein (2-79AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CKS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human