Anti-AKT3 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77849-591
- 77849-592
- Antigen symbol:
- AKT3
- AKT3
- Antigen synonyms:
- AKT3 kinase|AKT3_HUMAN|DKFZp434N0250|MPPH|PKB gamma|PKBG|PRKBG|Protein kinase Akt-3|Protein Kinase Protein kinase B gamma|RAC gamma|RAC gamma serine/threonine protein kinase|RAC-gamma serine/threonine-protein kinase|RAC-PK-gamma|RACPK Gamma|Serine threonine protein kinase Akt 3|Serine threonine protein kinase STK 2|STK-2|STK2|V akt murine thymoma viral oncogene homolog 3 (protein kinase B|gamma)|V akt murine thymoma viral oncogene homolog 3|V akt murine thymoma viral oncogene homolog 3 protein kinase B gamma
- AKT3 kinase|AKT3_HUMAN|DKFZp434N0250|MPPH|PKB gamma|PKBG|PRKBG|Protein kinase Akt-3|Protein Kinase Protein kinase B gamma|RAC gamma|RAC gamma serine/threonine protein kinase|RAC-gamma serine/threonine-protein kinase|RAC-PK-gamma|RACPK Gamma|Serine threonine protein kinase Akt 3|Serine threonine protein kinase STK 2|STK-2|STK2|V akt murine thymoma viral oncogene homolog 3 (protein kinase B|gamma)|V akt murine thymoma viral oncogene homolog 3|V akt murine thymoma viral oncogene homolog 3 protein kinase B gamma
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y243
- Q9Y243
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human AKT3
- Synthetic peptide of Human AKT3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together