Anti-ACTR1A Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-689
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ACTR1A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P61163
- Antigen synonyms:Actin related protein 1|Actin RPV|Actin-RPV|ACTR 1A|ACTZ_HUMAN|Alpha centractin|Alpha-centractin|ARP 1|ARP1 actin related protein 1 homolog A|ARP1 actin related protein 1 homolog A centractin alpha|ARP1|ARP1 yeast homolog A|Centractin alpha|Centractin|Centrosome associated actin homolog|Centrosome-associated actin homolog|CTRN 1|CTRN1|FLJ52695|FLJ52800|FLJ55002
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-centractin protein (187-376AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ACTR1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human