Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Mesothelin (from HEK293 cells)
Human Recombinant Mesothelin (from HEK293 cells)
Supplier:  Antibodies.com
Certificates
Human Recombinant Mesothelin (from HEK293 cells)
Supplier:  Antibodies.com

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Gene ID:
    UniprotID# Q13421
  • Reconstitution instructions:
    Prior to reconstitution, centrifuge the vial at 5000 g for 3 - 5 minutes at room temperature. Reconstitute with appropriate volume of distilled sterile water to bring product to 1 mg/ml concentration. After addition of distilled sterile water, mix by gentle tapping. Note: It is not recommended to vortex or vigorously pipette the sample.
  • Carrier-free:
    N
  • Protein synonyms:
    CAK1 antigen|MPF|MSLN|Pre-pro-megakaryocyte-potentiating factor
  • Protein/peptide name:
    Mesothelin
  • Purity:
    >95% by SDS-PAGE and coomassie blue staining
  • Molecular weight:
    The protein has a predicted molecular mass of 59.3 kDa after removal of the signal peptide
  • Sequence:
    Mesothelin (Glu296-Gly580) + mFc (Pro99-Lys330) + 6×His Tag
  • Concentration:
    Reconstitution dependent
  • Formulation:
    Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Tested applications:
    ELISA, SDS-PAGE

Specifications

Frequently Bought Together