Anti-PSG11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSG11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-382
- 77844-381
- Antigen symbol:
- PSG11
- PSG11
- Antigen synonyms:
- MGC22484|OTTHUMP00000200192|OTTHUMP00000200193|OTTHUMP00000200194|Pregnancy specific beta 1 glycoprotein 13|Pregnancy-specific beta-1-glycoprotein 11|Pregnancy-specific beta-1-glycoprotein 13|Pregnancy-specific glycoprotein 11|Pregnancy-specific glycoprotein 13|PS-beta-G-11|PS-beta-G-13|PSBG 11|PSBG 13|PSBG-11|PSBG-13|PSBG11|PSBG13|PSG 11|PSG11_HUMAN|PSG13|PSG14
- MGC22484|OTTHUMP00000200192|OTTHUMP00000200193|OTTHUMP00000200194|Pregnancy specific beta 1 glycoprotein 13|Pregnancy-specific beta-1-glycoprotein 11|Pregnancy-specific beta-1-glycoprotein 13|Pregnancy-specific glycoprotein 11|Pregnancy-specific glycoprotein 13|PS-beta-G-11|PS-beta-G-13|PSBG 11|PSBG 13|PSBG-11|PSBG-13|PSBG11|PSBG13|PSG 11|PSG11_HUMAN|PSG13|PSG14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQ72
- Q9UQ72
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Pregnancy-specific beta-1-glycoprotein 11 protein (7-219AA)
- Recombinant human Pregnancy-specific beta-1-glycoprotein 11 protein (7-219AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together