Anti-ADCY8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-768
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ADCY8
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P40145
- Antigen synonyms:Adenylate cyclase type 8|EC 4.6.1.1|ATP pyrophosphate-lyase 8|Adenylate cyclase type VIII|Adenylyl cyclase 8|AC8|Ca(2+)/calmodulin-activated adenylyl cyclase
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenylate cyclase type 8 protein (534-670AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ADCY8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human