Anti-TAF6L Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAF6L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-793
- 77845-792
- Antigen symbol:
- TAF6L
- TAF6L
- Antigen synonyms:
- PAF65A|TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L|PCAF-associated factor 65-alpha|PAF65-alpha
- PAF65A|TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L|PCAF-associated factor 65-alpha|PAF65-alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6J9
- Q9Y6J9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L protein (345-433AA)
- Recombinant Human TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L protein (345-433AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together