Order Entry
Canada
ContactUsLinkComponent
Human Recombinant Angiotensin Converting Enzyme-2 (from NS0 cells)
Human Recombinant Angiotensin Converting Enzyme-2 (from NS0 cells)
Catalog # CA80603-766
Supplier:  MilliporeSigma
CAS Number:  
Human Recombinant Angiotensin Converting Enzyme-2 (from NS0 cells)
Catalog # CA80603-766
Supplier:  MilliporeSigma
Supplier Number:  176872-10UG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Enzyme type:
    Recombinant
  • Source:
    NS0 cells
  • Species:
    Human
  • Size:
    10 μg
  • Storage conditions:
    Safe to freeze, avoid repeated freeze/thaw cycles
  • Enzyme name:
    Angiotensin Converting Enzyme-2
  • Enzyme synonyms:
    ACE Homolog;ACEH;ACE-2
  • Purity:
    ≥90% by SDS-PAGE

Specifications

About this item