Anti-ABHD2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-553
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ABHD2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P08910
- Antigen synonyms:LABH2Monoacylglycerol lipase EC 3.1.1.23|2-arachidonoylglycerol hydrolase|Abhydrolase domain-containing protein 2|Acetylesterase|EC 3.1.1.6|Lung alpha/beta hydrolase 2|Progesterone-sensitive lipase|EC 3.1.1.79|Protein PHPS1-2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Monoacylglycerol lipase ABHD2 protein (31-320AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human