Anti-CCR6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77852-575
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCR6
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P51684
- Antigen synonyms:BN 1|BN1|C C chemokine receptor type 6|C C CKR 6|C-C chemokine receptor type 6|C-C CKR-6|CC chemokine receptor type 6|CC CKR 6|CC R6|CC-CKR-6|CCCKR6|CCR 6|CCR-6|Ccr6|CCR6_HUMAN|CD 196|CD196|CD196 antigen|Chemokine (C C motif) receptor 6|Chemokine (C C) receptor 6|Chemokine (CC motif) receptor 6|Chemokine (CC) receptor 6|Chemokine receptor 6|Chemokine receptor like 3|Chemokine receptor-like 3|CKR 6|CKR L3|CKR-L3|CKR6|CKRL3|CMKBR 6|CMKBR6|DCR 2|DCR2|DRY 6|DRY6|G protein coupled receptor 29|G-protein coupled receptor 29|GPR 29|GPR CY4|GPR-CY4|GPR29|GPRCY 4|GPRCY4|LARC receptor|Seven transmembrane receptor lymphocyte 22|STRL 22|STRL22
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-C chemokine receptor type 6 protein (322-374AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCR6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human