Anti-HARS Rabbit Recombinant Polyclonal Antibody
Catalog # 77856-718
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HARS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P12081
- Antigen synonyms:cytoplasmic|EC 6.1.1.21|FLJ20491|HisRS|Histidine tRNA ligase, cytoplasmic|histidine translase|Histidine tRNA ligase|Histidine--tRNA ligase|Histidyl tRNA synthetase|Histidyl-tRNA synthetase|HRS|Human histidyl tRNA synthetase homolog (HO3) mRNA complete cds|SYHC_HUMAN|USH3B
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Histidine--tRNA ligase, cytoplasmic protein (1-180AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000, IHC:1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human