Anti-CHUK (phospho-Thr23) Rabbit Polyclonal Antibody
Catalog # 77847-366
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CHUK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:O15111
- Antigen synonyms:IkappaB kinase|Transcription factor 16|I-kappa-B kinase alpha|Inhibitor of nuclear factor kappa-B kinase subunit alpha|Nuclear factor NFkappaB inhibitor kinase alpha|chuk|Nuclear factor NF kappa B inhibitor kinase alpha|IKK 1|Conserved helix loop ubiquitous kinase|Nuclear Factor Of Kappa Light Chain Gene Enhancer In B Cells Inhibitor|IKKA_HUMAN|CHUK1|Conserved helix-loop-helix ubiquitous kinase|Nuclear factor NF-kappa-B inhibitor kinase alpha|TCF16|Conserved Helix Loop Helix Ubiquitous Kinase|IkBKA|IKK A|Nuclear Factor Kappa B Inhibitor Kinase Alpha|IKK-alpha|IKK1|I Kappa B Kinase Alpha|NFKBIKA|IkB kinase alpha subunit|TCF-16|I Kappa B Kinase 1|Inhibitor Of Nuclear Factor Kappa B Kinase Alpha Subunit|I-kappa-B kinase 1|IKKA|Inhibitor Of Kappa Light Polypeptide Gene Enhancer In B Cells|IKK a kinase|IKK-A
- Modification:phospho-Thr23
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of threonine 23 (L-G-T(p)-G-G) derived from Human IKK a.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:1000, IHC:1:50 to 1:100
Type:
Antigen: CHUK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat