Anti-RALY Rabbit Recombinant Polyclonal Antibody
77844-896
New Product
- Antigen symbol:RALY
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UKM9
- Antigen synonyms:Autoantigen p542|Heterogeneous nuclear ribonucleoprotein C-like 2|hnRNP associated with lethal yellow protein homolog|hnRNP core protein C-like 2|MGC117312|P542|RALY_HUMAN|RNA binding protein (autoantigenic hnRNP associated with lethal yellow)|RNA binding protein (autoantigenic)|RNA binding protein autoantigenic (hnRNP associated with lethal yellow homolog (mouse))|RNA-binding protein Raly
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human RNA-binding protein Raly protein (254-306AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200
Type:
Antigen: RALY
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human