Anti-Ctcfl Rabbit Recombinant Polyclonal Antibody
77853-712
New Product
- Antigen symbol:Ctcfl
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse,Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:A2APF3
- Antigen synonyms:Boris|Transcriptional repressor Brother of the regulator of imprinted sites|CCCTC-binding factor|CTCF paralog|CTCF-like protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Mouse Transcriptional repressor CTCFL protein (1-636AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: Ctcfl
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human