Anti-AP3S2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-231
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CSTF2T
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H0L4
- Antigen synonyms:CF 1 64 kDa subunit tau|CF-1 64 kDa subunit tau variant|CF1 64 kDa subunit|tau variant|Cleavage stimulation factor 3' pre RNA|subunit 2 64kDa tau variant|Cleavage stimulation factor 64 kDa subunit tau|Cleavage stimulation factor 64 kDa subunit tau variant|Cleavage stimulation factor 64 kDa subunit|Cleavage stimulation factor subunit 2 tau|Cleavage stimulation factor subunit 2 tau variant|CSTF 64 kDa subunit tau|CSTF 64 kDa subunit tau variant|CSTF 64 kDa subunit|CstF 64T|CSTF2T|CSTF2T f|CSTFT_HUMAN|KIAA0689|TauCstF-64|TauCstF64
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:CF 1 64 kDa subunit tau
- Purification:Antigen affinity purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human