Anti-ASGR2 Rabbit Recombinant polyclonal Antibody (Biotinylated)
77851-472
New Product
- Antigen symbol:ASGR2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P07307
- Antigen synonyms:ASGP R|ASGP-R 2|ASGPR 2|ASGPR2|ASGR2_HUMAN|Asialoglycoprotein receptor 2|C type lectin domain family 4 member H2|C-type lectin domain family 4 member H2|CLEC4H2|HBxAg-binding protein|HBXBP|Hepatic lectin 2|Hepatic lectin H2|Hepatic lectin R2/3|HL-2|Hs.1259|L H2|LH2|MGC108546|MHL 2|RHL2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Asialoglycoprotein receptor 2 protein (63-286AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
0
Type:
Antigen: ASGR2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human