Order Entry
Canada
ContactUsLinkComponent
Anti-CAP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-CAP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-322
Supplier:  AFG Bioscience
New Product
undefined
Anti-CAP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-322
Supplier:  AFG Bioscience
Supplier Number:  A15895-50UG
New Product

Specifications

  • Antigen symbol:
    CAP2
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    P40123
  • Antigen synonyms:
    2810452G09Rik|adenylate cyclase associated protein 2|Adenylyl cyclase associated protein 2|Adenylyl cyclase-associated protein 2|CAP 2|CAP adenylate cyclase associated protein 2|CAP adenylate cyclase-associated protein 2|CAP|adenylate cyclase associated protein|2 (yeast)|cap2|CAP2_HUMAN
  • Storage buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Adenylyl cyclase-associated protein 2 protein (221-477AA)
  • Purification:
    >95% Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: CAP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human