Anti-MMACHC Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MMACHC
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-895
- 77845-896
- Antigen symbol:
- MMACHC
- MMACHC
- Antigen synonyms:
- 1810037K07Rik|BOS_3654|cblC|DKFZp564I122|FLJ25671|Methylmalonic aciduria (cobalamin deficiency) cblC type with homocystinuria|Methylmalonic aciduria and homocystinuria type C protein|Methylmalonic aciduria and homocystinuria type C protein homolog|MGC134307|MMAC_HUMAN|OTTHUMP00000009243|RP11 291L19.3|RP23-177C18.3
- 1810037K07Rik|BOS_3654|cblC|DKFZp564I122|FLJ25671|Methylmalonic aciduria (cobalamin deficiency) cblC type with homocystinuria|Methylmalonic aciduria and homocystinuria type C protein|Methylmalonic aciduria and homocystinuria type C protein homolog|MGC134307|MMAC_HUMAN|OTTHUMP00000009243|RP11 291L19.3|RP23-177C18.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4U1
- Q9Y4U1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Methylmalonic aciduria and homocystinuria type C protein (63-282AA)
- Recombinant Human Methylmalonic aciduria and homocystinuria type C protein (63-282AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together