Anti-CD160 Rabbit Recombinant Polyclonal Antibody
77852-609
New Product
- Antigen symbol:CD160
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95971
- Antigen synonyms:BY55|BY55_HUMAN|CD160 antigen [Precursor]|CD160 antigen|CD160 delta Ig|CD160 molecule|CD160 transmembrane isoform|FLJ46513|Natural killer cell receptor BY55|Natural killer cell receptor|immunoglobulin superfamily member|NK1|NK28
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human CD160 antigen protein (27-159AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CD160
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human