Anti-CCT5 Rabbit Recombinant Polyclonal Antibody
77852-595
New Product
- Antigen symbol:CCT5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P48643
- Antigen synonyms:CCT 5| CCT epsilon| CCT-epsilon| CCT5| CCTE| CCTepsilon| Chaperonin containing T-complex polypeptide 1| subunit 5| Chaperonin containing TCP 1 epsilon| Chaperonin containing TCP 1 subunit 5| Chaperonin containing TCP 1 subunit 5 epsilon| Chaperonin containing TCP1 epsilon| Chaperonin containing TCP1 subunit 5| Chaperonin containing TCP1 subunit 5 epsilon| KIAA0098| T complex protein 1 epsilon subunit| T complex protein 1 subunit epsilon| T-complex protein 1 subunit epsilon| TCP 1 epsilon| TCP-1-epsilon| TCP1epsilon| TCPE| TCPE_HUMAN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-complex protein 1 subunit epsilon protein (1-270AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CCT5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse