Anti-GTPBP4 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GTPBP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-015
- 77843-016
- Antigen symbol:
- GTPBP4
- GTPBP4
- Antigen synonyms:
- Chronic renal failure gene protein|CRFG|G protein binding protein CRFG|GTP binding protein 4|GTP-binding protein NGB|NGB|NOG1|NOG1_HUMAN|Nucleolar GTP binding protein 1|Nucleolar GTP-binding protein 1
- Chronic renal failure gene protein|CRFG|G protein binding protein CRFG|GTP binding protein 4|GTP-binding protein NGB|NGB|NOG1|NOG1_HUMAN|Nucleolar GTP binding protein 1|Nucleolar GTP-binding protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZE4
- Q9BZE4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Nucleolar GTP-binding protein 1 protein (347-634AA)
- Recombinant Human Nucleolar GTP-binding protein 1 protein (347-634AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together