Anti-RPSA Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:100 to 1:300
Type:
Antigen: RPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77848-083
- 77848-082
- Antigen symbol:
- RPSA
- RPSA
- Antigen synonyms:
- 34/67 kDa laminin receptor|37 kDa laminin receptor precursor|37/67 kDa laminin receptor|37LRP|40S ribosomal protein SA|67 kDa laminin receptor|67LR|Colon carcinoma laminin binding protein|Colon carcinoma laminin-binding protein|LAMBR|Laminin receptor 1|Laminin-binding protein precursor p40|LAMR 1|LamR|LAMR1|LBP|LBP/p40|LRP|LRP/LR|Multidrug resistance associated protein MGr1 Ag|Multidrug resistance associated protein MGr1Ag|Multidrug resistance-associated protein MGr1-Ag|NEM/1CHD4|p40|Ribosomal Protein SA|rpsA|RSSA_HUMAN|SA
- 34/67 kDa laminin receptor|37 kDa laminin receptor precursor|37/67 kDa laminin receptor|37LRP|40S ribosomal protein SA|67 kDa laminin receptor|67LR|Colon carcinoma laminin binding protein|Colon carcinoma laminin-binding protein|LAMBR|Laminin receptor 1|Laminin-binding protein precursor p40|LAMR 1|LamR|LAMR1|LBP|LBP/p40|LRP|LRP/LR|Multidrug resistance associated protein MGr1 Ag|Multidrug resistance associated protein MGr1Ag|Multidrug resistance-associated protein MGr1-Ag|NEM/1CHD4|p40|Ribosomal Protein SA|rpsA|RSSA_HUMAN|SA
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08865
- P08865
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human RPSA
- Fusion protein of Human RPSA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together