Anti-MAPK7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:30 to 1:150
Type:
Antigen: MAPK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77846-945
- 77846-946
- Antigen symbol:
- MAPK7
- MAPK7
- Antigen synonyms:
- Big MAP kinase 1|BMK 1|BMK 1 kinase|BMK-1|BMK1|BMK1 Kinase|EC 2.7.11.24|ERK 4|ERK 5|ERK-5|ERK4|ERK5|Extracellular signal regulated kinase 5|Extracellular signal-regulated kinase 5|MAP kinase 7|MAPK 7|MAPK7|Mitogen activated protein kinase 7|Mitogen-activated protein kinase 7|MK07_HUMAN|OTTHUMP00000065906|OTTHUMP00000065907|PRKM 7|PRKM7|PROTEIN KINASE|MITOGEN-ACTIVATED|7
- Big MAP kinase 1|BMK 1|BMK 1 kinase|BMK-1|BMK1|BMK1 Kinase|EC 2.7.11.24|ERK 4|ERK 5|ERK-5|ERK4|ERK5|Extracellular signal regulated kinase 5|Extracellular signal-regulated kinase 5|MAP kinase 7|MAPK 7|MAPK7|Mitogen activated protein kinase 7|Mitogen-activated protein kinase 7|MK07_HUMAN|OTTHUMP00000065906|OTTHUMP00000065907|PRKM 7|PRKM7|PROTEIN KINASE|MITOGEN-ACTIVATED|7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13164
- Q13164
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MAPK7
- Fusion protein of Human MAPK7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together