Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human Prothrombin (from Plasma)
Human Prothrombin (from Plasma)
:
MilliporeSigma
Human Prothrombin (from Plasma)
:
MilliporeSigma
CAS No.:
9001-26-7
Source:
Plasma
Species:
Human
Storage conditions:
Safe to freeze
Protein synonyms:
Coagulation Factor II
Protein/peptide name:
Prothrombin
Purity:
>95% (SDS-PAGE)
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning