Recombinant Human CD55 (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
Preserved Starfish
Supplier: Avantor
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Buffers, Precision Reference Standards, RICCA Chemical Company
Supplier: Ricca Chemical
Fluoride Standard, 1 ml = 0.1 mg F-, 100 ppm F-, Ricca Chemical Company
Supplier: Ricca Chemical
Reagent Sets for Colorimeters, Hach
Supplier: Hach
ClearSteel, Stainless Steel Cleaner and Polish Spray, Mortech
Supplier: Mortech Manufacturing
Iron(III) chloride hexahydrate 97%
Supplier: Thermo Scientific Chemicals
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CIP 100® Alkaline Process and Research Cleaners
Supplier: Steris
Human Recombinant PDGF-AA (from <i>E. coli</i>)
Supplier: Peprotech