Anti-FEN1 Rabbit Polyclonal Antibody
Catalog # 77847-695
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FEN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P39748
- Antigen synonyms:DNase IV| FEN-1| FEN1_HUMAN| Flap endonuclease 1| Flap structure specific endonuclease 1| Flap structure-specific endonuclease 1| hFEN-1| h Maturation factor 1| MF1| Rad2
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Synthesized peptide derived from internal of Human FEN1.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500
Type:
Antigen: FEN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat