Anti-EIF4EBP2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77854-627
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:EIF4EBP2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q13542
- Antigen synonyms:4E-BP2|4EBP2|4EBP2_HUMAN|eIF4E binding protein 2|eIF4E-binding protein 2|Eukaryotic translation initiation factor 4E binding protein 1|Eukaryotic translation initiation factor 4E-binding protein 2|PHASII|phosphorylated|heat and acid stable regulated by insulin protein II
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Eukaryotic translation initiation factor 4E-binding protein 2 protein (1-120AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EIF4EBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human