Order Entry
Canada
ContactUsLinkComponent
Anti-ABI2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-ABI2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-564
Supplier:  AFG Bioscience
New Product
undefined
Anti-ABI2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77850-564
Supplier:  AFG Bioscience
Supplier Number:  A11613-50UG
New Product

Specifications

  • Antigen symbol:
    ABI2
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    Q9NYB9
  • Antigen synonyms:
    Abelson interactor 2|ABI 2|Abi-2|ABI2|ABI2_HUMAN|ABI2B|Abl binding protein 3|Abl interacting protein 1 (SH3 containing protein)|Abl interactor 2|Abl interactor protein 2b|Abl-binding protein 3|AblBP3|AIP 1|Arg binding protein 1|Arg protein tyrosine kinase binding protein|Arg-binding protein 1|ArgBP1|ARGBPIA|ArgBPIB|OTTHUMP00000163783|OTTHUMP00000163784|OTTHUMP00000206182|OTTHUMP00000206183|OTTHUMP00000206184|OTTHUMP00000206185|SSH3BP2
  • Storage buffer:
    0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Abl interactor 2 protein (164-452AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: ABI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human