Anti-ACTL6B Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-679
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ACTL6B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O94805
- Antigen synonyms:53 kDa BRG1 associated factor B|53 kDa BRG1-associated factor B|ACL 6B|ACL6B|ACL6B_HUMAN|Actin like 6|Actin like protein 6B|Actin like protein6B|Actin related protein|Actin related protein Baf53b|actin-like 6B|Actin-like protein 6B|Actin-related protein Baf53b|Actl 6|ACTL 6B|Actl6|ACTL6 B|Actl6b|Arp Na|ArpN a|ArpN alpha|ArpNa|ArpNalpha|Baf 53b|BAF53B|BRG1-associated factor 53B
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Actin-like protein 6B protein (1-280AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ACTL6B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human