Anti-MNX1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-862
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MNX1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P50219
- Antigen synonyms:HB 9|HB9|HLXB 9|HLXB9|Homeo box HB9|Homeobox HB9|Homeobox protein HB9|HOXHB9|MNX1_HUMAN|Motor neuron and pancreas homeobox 1|Motor neuron and pancreas homeobox protein 1|Sacral agenesis autosomal dominant (Currarino triad)|SCRA 1|SCRA1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Motor neuron and pancreas homeobox protein 1 protein (50-99AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: MNX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human