Anti-CDH1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CDH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77847-074
- 77847-075
- Antigen symbol:
- CDH1
- CDH1
- Antigen synonyms:
- Arc 1| CADH1_HUMAN| Cadherin 1| cadherin 1 type 1 E-cadherin| Cadherin-1| Cadherin1| CAM 120/80| CD 324| CD324| CD324 antigen| cdh1| CDHE| E-Cad/CTF3| E-cadherin| ECAD| Epithelial cadherin| epithelial calcium dependant adhesion protein| LCAM| Liver cell adhesion molecule| UVO| Uvomorulin
- Arc 1| CADH1_HUMAN| Cadherin 1| cadherin 1 type 1 E-cadherin| Cadherin-1| Cadherin1| CAM 120/80| CD 324| CD324| CD324 antigen| cdh1| CDHE| E-Cad/CTF3| E-cadherin| ECAD| Epithelial cadherin| epithelial calcium dependant adhesion protein| LCAM| Liver cell adhesion molecule| UVO| Uvomorulin
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12830
- P12830
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human pan CDH
- Synthetic peptide of Human pan CDH
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together