Anti-ABAT Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-503
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ABAT
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P80404
- Antigen synonyms:(S) 3 amino 2 methylpropionate transaminase|(S)-3-amino-2-methylpropionate transaminase|4 aminobutyrate aminotransferase|mitochondrial|4-aminobutyrate aminotransferase|FLJ17813|FLJ30272|GABA aminotransferase|GABA AT|GABA T|GABA transaminase|GABA transferase|GABA-AT|GABA-T|GGABT_HUMAN|Gamma amino N butyrate transaminase|Gamma-amino-N-butyrate transaminase|hCG1984265|L AIBAT|L-AIBAT|LAIBAT|NPD009
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human 4-aminobutyrate aminotransferase, mitochondrial protein (29-300AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ABAT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat