Anti-HRH3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HRH3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-584
- 77846-585
- Antigen symbol:
- HRH3
- HRH3
- Antigen synonyms:
- G protein coupled receptor 97|G-protein coupled receptor 97|GPCR 97|GPCR97|H3R|HH 3R|HH3 R|HH3R|Histamine H3 receptor|Histamine receptor H3|HRH 3|HRH3|HRH3_HUMAN|OTTHUMP00000031459
- G protein coupled receptor 97|G-protein coupled receptor 97|GPCR 97|GPCR97|H3R|HH 3R|HH3 R|HH3R|Histamine H3 receptor|Histamine receptor H3|HRH 3|HRH3|HRH3_HUMAN|OTTHUMP00000031459
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5N1
- Q9Y5N1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histamine H3 receptor protein (236-332AA)
- Recombinant Human Histamine H3 receptor protein (236-332AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together