Anti-KLF13 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: KLF13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-332
- 77846-333
- Antigen symbol:
- KLF13
- KLF13
- Antigen synonyms:
- Basic transcription element binding protein 3|Basic transcription element-binding protein 3|BTE binding protein 3|BTE-binding protein 3|BTEB3|C2 H2 zinc finger protein|FKLF 2|FKLF2|Klf13|KLF13_HUMAN|Krueppel-like factor 13|Kruppel like factor 13|Novel Sp1 like zinc finger transcription factor |Novel Sp1-like zinc finger transcription factor 1|NSLP1|RANTES factor of late activated T lymphocytes 1|RANTES factor of late activated T-lymphocytes 1|RFLAT 1|RFLAT-1|RFLAT1|Transcription factor BTEB3|Transcription factor NSLP1
- Basic transcription element binding protein 3|Basic transcription element-binding protein 3|BTE binding protein 3|BTE-binding protein 3|BTEB3|C2 H2 zinc finger protein|FKLF 2|FKLF2|Klf13|KLF13_HUMAN|Krueppel-like factor 13|Kruppel like factor 13|Novel Sp1 like zinc finger transcription factor |Novel Sp1-like zinc finger transcription factor 1|NSLP1|RANTES factor of late activated T lymphocytes 1|RANTES factor of late activated T-lymphocytes 1|RFLAT 1|RFLAT-1|RFLAT1|Transcription factor BTEB3|Transcription factor NSLP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2Y9
- Q9Y2Y9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Krueppel-like factor 13 protein (26-147AA)
- Recombinant Human Krueppel-like factor 13 protein (26-147AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together