Anti-CCR2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-554
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCR2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P41597
- Antigen synonyms:C C chemokine receptor type 2|C C CKR 2|C-C chemokine receptor type 2|C-C CKR-2|CC chemokine receptor type 2|CC CKR 2|CC-CKR-2|CCCKR2|CCR 2|CCR-2|CCR1L|CCR2_HUMAN|CCR2A|CCR2B|CCR5L|CD192|CD192 antigen|Chemokine C C motif receptor 2|Chemokine CC Motif Receptor 2|CKR 2|CKR2|CKR2A|CKR2B|CMKBR2|MCP 1 R|MCP-1-R|MCP1 RECEPTOR|MCP1R|Monocyte chemoattractant protein 1 receptor|Monocyte Chemotactic Protein 1 Receptor
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-C chemokine receptor type 2 protein (310-374AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human