Order Entry
Canada
ContactUsLinkComponent
Anti-EDAR Rabbit Recombinant Polyclonal Antibody (Biotin)
Anti-EDAR Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-EDAR Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77845-011
  • 77845-012
  • Antigen symbol:
  • EDAR
  • EDAR
  • Antigen synonyms:
  • Anhidrotic ectodysplasin receptor 1|DL|Downless (mouse) homolog|Downless homolog|Downless mouse homolog of|ECTD10A|ECTD10B|Ectodermal dysplasia receptor|Ectodysplasin 1 anhidrotic receptor|Ectodysplasin A receptor|Ectodysplasin A1 isoform receptor|Ectodysplasin-A receptor|ED 1R|ED 3|ED 5|ED1R|ED3|ED5|EDA 1R|EDA 3|EDA A1 receptor|EDA A1R|EDA-A1 receptor|EDA1R|EDA3|Edar|EDAR_HUMAN|HRM1|Tumor necrosis factor receptor superfamily member EDAR
  • Anhidrotic ectodysplasin receptor 1|DL|Downless (mouse) homolog|Downless homolog|Downless mouse homolog of|ECTD10A|ECTD10B|Ectodermal dysplasia receptor|Ectodysplasin 1 anhidrotic receptor|Ectodysplasin A receptor|Ectodysplasin A1 isoform receptor|Ectodysplasin-A receptor|ED 1R|ED 3|ED 5|ED1R|ED3|ED5|EDA 1R|EDA 3|EDA A1 receptor|EDA A1R|EDA-A1 receptor|EDA1R|EDA3|Edar|EDAR_HUMAN|HRM1|Tumor necrosis factor receptor superfamily member EDAR
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UNE0
  • Q9UNE0
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Tumor necrosis factor receptor superfamily member EDAR protein (27-170AA)
  • Recombinant Human Tumor necrosis factor receptor superfamily member EDAR protein (27-170AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95% Protein G purified
  • >95% Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together