Anti-ADAMTS18 Rabbit Polyclonal Antibody
Catalog # 77850-108
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ADAMTS18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8TE60
- Antigen synonyms:ADAMTS21A disintegrin and metalloproteinase with thrombospondin motifs 18|ADAM-TS 18|ADAM-TS18|ADAMTS-18|EC 3.4.24.-
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human ADAMTS18
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ADAMTS18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse