Anti-ADPGK Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-818
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ADPGK
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:Q9BRR6
- Antigen synonyms:2610017G09Rik|ADP dependent glucokinase|ADP GK|ADP-dependent glucokinase|ADP-GK|ADPGK_HUMAN|ATP dependent glucokinase|DKFZP434B195|RbBP 35|RbBP-35
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human ADP-dependent glucokinase protein (218-497AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ADPGK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human