Anti-ATE1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-516
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ATE1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95260
- Antigen synonyms:AI225793|Arginine tRNA protein transferase 1|Arginine-tRNA--protein transferase 1|Arginyl-tRNA--protein transferase 1|Arginyl-tRNA-protein transferase|Arginyltransferase 1|Ate1|ATE1_HUMAN|AW547406|EC 2.3.2.8|MGC26724|OTTHUMP00000020637|OTTHUMP00000058665|R transferase 1|R-transferase 1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Arginyl-tRNA--protein transferase 1 protein (105-253AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human